Principaux documents
SYNT230012
Synthware graduated distillation receiver tube with hooks
capacity 12 mL, joint: ST/NS 14/20
Synonyme(s) :
Synthware T230012, distilling receiver tube
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.51
Matériaux
V-bottom bottom
colorless tube
glass tube
Joint
joint: ST/NS14/20
Caractéristiques
graduations
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware T230012
Capacité
12 mL
Description générale
Centrifuge tube shaped receiver with an upper standard taper joint.
Informations légales
Synthware is a trademark of Kemtech America Inc.