Principaux documents
SYNP150500M
Synthware synthesis vessel with lower joint and PTFE T-bore stopcock
500 mL, joint: ST/NS 24/40, porosity: medium, with drip tip
Synonyme(s) :
Synthware P150500M, Solid phase peptide synthesis vessel
About This Item
Code UNSPSC :
41100000
Nomenclature NACRES :
NB.51
Joint
joint: ST/NS24/40
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware P150500M
Capacité
500 mL
500 mL
Description générale
Solid phase peptide synthesis vessels feature 2 mm PTFE stopcocks and fritted glass supports for resin beads during operation. The 24/40 inner drip joint can be assembled with other standard glassware. The vessel has a 8 mm hose connection for vacuum and the stopcock has an 8 mm O.D. sidearm for inert gas introduction. Vessels are supplied with PTFE-faced silicone lined Nylon cap.
Informations légales
Synthware is a trademark of Kemtech America Inc.