Key Documents
SYNF966024
SynthwareBuchner funnel with perforated plate
150 mL, joint: ST/NS 24/40, plate O.D. 60 mm
Synonyme(s) :
Synthware F966024, filter funnel
About This Item
Code UNSPSC :
41121809
Nomenclature NACRES :
NB.51
Joint
joint: ST/NS24/40
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware F966024
Capacité
150 mL
150 mL
D.E. de la plaque/du plateau
60 mm
Informations légales
Synthware is a trademark of Kemtech America Inc.