Principaux documents
SYNF904100H
Synthware pear-shaped Schlenk flask with stopcock
capacity 100 mL, joint: ST/NS 24/40
Synonyme(s) :
Synthware F904100H, modified Schlenk reaction flask
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.51
Matériaux
borosilicate glass flask
pear-shaped
single-neck
Joint
joint: ST/NS24/40
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware F904100H
Capacité
100 mL
Description générale
A suffix of G indicates a glass stopcock, H indicates a high-vacuum valve, no letter suffix indicates a PTFE Stopcock.
Informations légales
Synthware is a trademark of Kemtech America Inc.