Principaux documents
SYND201414
Synthware vertical distilling adapter
Cow Receiver Only, joint: ST/NS 14/20
Synonyme(s) :
Synthware D201414, cow type distilling receiver
About This Item
Code UNSPSC :
41104812
Nomenclature NACRES :
NB.51
Joint
joint: ST/NS14/20
Caractéristiques
Cow Receiver Only
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware D201414
Description générale
Designed without hose connection to be used with D10 distillation apparatus.
Informations légales
Synthware is a trademark of Kemtech America Inc.