Principaux documents
SYNC366040C
Synthware chromatography column with PTFE stopcock and coarse frit
L 457 mm
Synonyme(s) :
Synthware C366040C, glass chromatograpy column
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.51
Qualité
for chromatography
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware C366040C
Longueur
457 mm
Description générale
With reinforced beaded top, coarse porosity fritted disc to support column packing. All Synthware columns are heavy wall designed to increase safety.
Informations légales
Synthware is a trademark of Kemtech America Inc.