Changer de vue
| À vous/Référence | Disponibilité | Prix |
|---|
A propos de cet article
NACRES:
NB.51
UNSPSC Code:
41121800
Manufacturer/tradename:
Synthware C174200
Packaging:
pkg of 1 ea
Joint:
joint: ST/NS24/40
Le tarif et la disponibilité ne sont pas disponibles actuellement.
Service technique
Besoin d'aide ? Notre équipe de scientifiques expérimentés est là pour vous.
Laissez-nous vous aiderjoint
joint: ST/NS24/40
packaging
pkg of 1 ea
manufacturer/tradename
Synthware C174200
jacket H
200 mm
General description
Indentations at the bottom for supporting packing material. Jacket permits the column to be used as a condenser.
Legal Information
Synthware is a trademark of Kemtech America Inc.
1 of 1
Cet article | |||
|---|---|---|---|
| description joint: ST/NS 24/40, jacket H 200 mm | description joint: ST/NS 14/20, jacket H 110 mm | description joint: ST/NS 14/20, jacket H 200 mm | description joint: ST/NS 19/22, jacket H 110 mm |
| manufacturer/tradename Synthware C174200 | manufacturer/tradename Synthware C171120 | manufacturer/tradename Synthware C171200 | manufacturer/tradename Synthware C179110 |
| joint joint: ST/NS24/40 | joint joint: ST/NS14/20 | joint joint: ST/NS14/20 | joint joint: ST/NS19/22 |
| packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea |
| jacket H 200 mm | jacket H 110 mm | jacket H 200 mm | jacket H 110 mm |
Still not finding the right product?
Explore all of our products under Synthware™ water jacketed distillation column with indents to support packing material