Principaux documents
SYNA372942
Synthware Claisen adapter
About This Item
Code UNSPSC :
41121815
Nomenclature NACRES :
NB.51
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware A372942
Description générale
Provides dual entry into a variety of flasks.
Informations légales
Synthware is a trademark of Kemtech America Inc.