Principaux documents
SYNA322440
Synthware vacuum adapter
female joint: ST/NS 24/40
Synonyme(s) :
Synthware A322440, glass vacuum adapter
About This Item
Code UNSPSC :
41121815
Nomenclature NACRES :
NB.51
Joint
female joint: ST/NS24/40
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware A322440
Description générale
Ideal for use as a vent or for the introduction of pressure/vacuum.
Informations légales
Synthware is a trademark of Kemtech America Inc.