Key Documents
SLW5810/4
Pyrex® Vacuum filtration system for 47 mm membranes
conical flask only
Synonyme(s) :
Pyrex, SciLabware 5810/4
About This Item
Code UNSPSC :
41104900
Nomenclature NACRES :
NB.43
Stérilité
non-sterile
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
5810/4
SciLabware
Description générale
Parts for vacuum filtration system for 47 mm membranes
  • Fully autoclavable
  • Ideal for microbiology
Informations légales
PYREX is a registered trademark of Corning, Inc.