Principaux documents
NOR5X4INSB
Optimizer insert for spinner turbines
NMR tube size 4 mm , into 5 mm Bruker
About This Item
Code UNSPSC :
41121705
Nomenclature NACRES :
SB.32
Matériaux
(acetal homopolymer resin)
Fabricant/nom de marque
Norell
Taille du tube de RMN
4 mm , into 5 mm Bruker
Description générale
Precision adapter holds 3 mm, 4 mm, and 4.25 mm NMR tube in a 5 mm spinner turbine. Does not include spinner turbine.