Principaux documents
BR458022
BRAND® filter crucible
borosilicate glass, capacity 30 mL, 1 D 2
Synonyme(s) :
Filter, Filter crucible
About This Item
Code UNSPSC :
41122200
Nomenclature NACRES :
NB.43
Matériaux
borosilicate glass
Fabricant/nom de marque
BRAND 458022
Capacité
30 mL
Dimension de pores
40-100 μm
Description générale
The BRAND® Filter Crucible made from Boro 3.3 glass is an essential tool for efficient filtration and separation processes in laboratory applications.
Informations légales
BRAND is a registered trademark of BRAND GMBH + CO KG