Principaux documents
BR452010
BRAND® disposable petri dish with lid
L × W 55 mm × 16 mm, vented
Synonyme(s) :
Dishes, Petri Dishes, Plastics
About This Item
Code UNSPSC :
41161502
Nomenclature NACRES :
NB.14
Matériaux
polystyrene
Caractéristiques
vented
Fabricant/nom de marque
BRAND 452010
L × l
55 mm × 16 mm
Description générale
Made from high-quality polystyrene (PS), these transparent dishes lids provide optimal visibility for observing cultures. Dishes and lids are robotically assembled to significantly reduce the risk of contamination, ensuring the integrity of your samples.
Caractéristiques et avantages
  • BRAND® disposable petri dish is manufactured from excellent quality polystyrene for sample clarity and visibility.
  • The transparent dishes and lids are assembled robotically to reduce the risk of contamination.
  • Able to withstand temperature up to 80 °C.
  • Fully stackable.
Informations légales
BRAND is a registered trademark of BRAND GMBH + CO KG