Principaux documents
BR131100
BRAND® stopcock for storage / aspirator bottles and drums
polypropylene
Synonyme(s) :
Stopcocks
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.35
Matériaux
polypropylene
Conditionnement
pack of 1 ea
Fabricant/nom de marque
BRAND 131100
Taille des connexions
3/4 in.
Description générale
The BRAND® stopcock for storage and aspirator bottles and drums is designed to fit all sizes, providing a versatile solution for fluid management in laboratory settings. It features a polypropylene (PP) adapter, ensuring compatibility with various containers. The stopcock comes with two FKM seals
Informations légales
BRAND is a registered trademark of BRAND GMBH + CO KG