Principaux documents
BAH195620000
SP Bel-Art® Straight Tubing Connectors
tubing I.D. 0.5-0.5 in. (6-12.7 mm)
Synonyme(s) :
connectors, tubing, tubing coupler
About This Item
Code UNSPSC :
41122800
Nomenclature NACRES :
NB.43
Conditionnement
pack of 12 ea
Fabricant/nom de marque
Bel-Art H19562-0000
L × largeur
1.75 in. (44.5 mm) × 0.5625 in. (14.3 mm)
D.I. du tuyau
0.5-0.5 in. (6-12.7 mm)
Description générale
These straight fittings ensure a tight, secure connection between lengths of tubing having the same inner diameter (I.D.).
Caractéristiques et avantages
  • Chemical-resistant polypropylene plastic for use with a wide range of liquids or gases.
  • FDA-grade for direct contact with food, drugs, and cosmetics
  • Autoclavable
Informations légales
Bel-Art is a registered trademark of SP Scienceware, a division of SP