Principaux documents
BAF131930500
Poxygrid® benchtop bag holder kit
includes 1 bag holder, 1 cover & 100 bags
Synonyme(s) :
bag holder
About This Item
Code UNSPSC :
41122800
Nomenclature NACRES :
NB.43
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Bel-Art F13193-0500
Description générale
Economical and convenient, this benchtop disposal system is perfect for collecting pipette tips, microcentrifuge tubes, etc. during daily use.
Caractéristiques et avantages
Economical and convenient, this benchtop disposal system is perfect for collecting pipette tips, microcentrifuge tubes, etc. during daily use.
  • Rectangular steel wire frame holds a polyethylene bag 22 x 28cm (8¹/₂ x 11"); coated in easy to clean, durable, orange color epoxy
  • Snug fitting polypropylene plastic cover helps hold the bag securely in place
  • Reusable; can be cleaned by hand or in automated dish washers; autoclavable
  • Cover provides compliance with 29 CFR 1910.1030(d)(2)(xiii)(A), which requires a cover on any container used to store, transport, or ship biohazardous wastes
  • Kit includes 100 Bench Top Biohazard Bags (F13166-0000); can also be used with Clavies Autoclavable Bags (H13182-0812)
Informations légales
Poxygrid is a registered trademark of SP Scienceware, a division of SP