Merck
  • Secreted IgM deficiency leads to increased BCR signaling that results in abnormal splenic B cell development.
Secreted IgM deficiency leads to increased BCR signaling that results in abnormal splenic B cell development.
Scientific reports (2017-06-16)
Dimitrios Tsiantoulas, Mate Kiss, Barbara Bartolini-Gritti, Andreas Bergthaler, Ziad Mallat, Hassan Jumaa, Christoph J Binder
ABSTRACT
Mice lacking secreted IgM (sIgM -/-) antibodies display abnormal splenic B cell development, which results in increased marginal zone and decreased follicular B cell numbers. However, the mechanism by which sIgM exhibit this effect is unknown. Here, we demonstrate that B cells in sIgM -/- mice display increased B cell receptor (BCR) signaling as judged by increased levels of phosphorylated Bruton's tyrosine kinase (pBtk), phosphorylated Spleen tyrosine kinase (pSyk), and nuclear receptor Nur77. Low dosage treatment with the pBtk inhibitor Ibrutinib reversed the altered B cell development in the spleen of sIgM -/- mice, suggesting that sIgM regulate splenic B cell differentiation by decreasing BCR signaling. Mechanistically, we show that B cells, which express BCRs specific to hen egg lysozyme (HEL) display diminished responsiveness to HEL stimulation in presence of soluble anti-HEL IgM antibodies. Our data identify sIgM as negative regulators of BCR signaling and suggest that they can act as decoy receptors for self-antigens that are recognized by membrane bound BCRs.
Sigma-Aldrich
Anti-Mouse IgM (μ-chain specific)–Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous glycerol solution
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-Mouse IgM (μ-chain specific) antibody produced in goat, affinity isolated antibody, lyophilized powder