Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C17H18N2O
CAS Number:
Molecular Weight:
266.34
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
eCl@ss:
25140904
grade
pharmaceutical primary standard
agency
USP
API family
tropicamide
manufacturer/tradename
USP
application(s)
pharmaceutical (small molecule)
pharmaceutical
pharmaceutical
format
neat
storage temp.
2-8°C
SMILES string
N(CC)(Cc2ccncc2)C(=O)C(=C)c1ccccc1
InChI
1S/C17H18N2O/c1-3-19(13-15-9-11-18-12-10-15)17(20)14(2)16-7-5-4-6-8-16/h4-12H,2-3,13H2,1H3
InChI key
LOVSQYAVWGMIRV-UHFFFAOYSA-N
General description
Find answers to frequently asked questions on USP products in USP FAQs.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.
Application
Tropicamide Related Compound B USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.
Also, for use with USP monograph such as
Tropicamide
Also, for use with USP monograph such as
Tropicamide
Analysis Note
These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.
Storage Class
10 - Combustible liquids
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.