Skip to Content
Merck

1486004

USP

Oxymetazoline hydrochloride

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

2-(3-Hydroxy-2,6-dimethyl-4-t-butylbenzyl)-2-imidazoline hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C16H24N2O · HCl
CAS Number:
Molecular Weight:
296.84
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

oxymetazoline

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Cl.Cc1cc(c(O)c(C)c1CC2=NCCN2)C(C)(C)C

InChI

1S/C16H24N2O.ClH/c1-10-8-13(16(3,4)5)15(19)11(2)12(10)9-14-17-6-7-18-14;/h8,19H,6-7,9H2,1-5H3,(H,17,18);1H

InChI key

BEEDODBODQVSIM-UHFFFAOYSA-N

General description

Find answers to frequently asked questions on USP products in USP FAQs.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Oxymetazoline hydrochloride USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Oxymetazoline Hydrochloride Ophthalmic Solution
  • Oxymetazoline Hydrochloride Nasal Solution

Biochem/physiol Actions

Oxymetazoline hydrochloride is a partial α2A-adrenoceptor agonist; agonist at 5-HT1A, 5-HT1B, and 5-HT1D serotonin receptors. It is a mixed agonist-antagonist at 5-HT1C serotonin receptors.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Disclaimer

Sales restrictions may apply.


Still not finding the right product?

Explore all of our products under Oxymetazoline hydrochloride


pictograms

Skull and crossbonesCorrosion

signalword

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 1 Oral - Aquatic Chronic 3 - Eye Dam. 1

Storage Class

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library