Skip to Content

1025602

USP

Amino methacrylate copolymer

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

2-Methyl-2-propenoic acid butyl ester polymer with 2-(dimethylamino)ethyl 2-methyl-2-propenoate and methyl 2-methyl-2-propenoate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
41116107
NACRES:
NA.24
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

amino methacrylate copolymer, basic butylated methacrylate copolymer, copolymers

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

N(CCOC(=O)C(=C)C)(C)C.O(CCCC)C(=O)C(=C)C.O(C)C(=O)C(=C)C

InChI

1S/C8H15NO2.C8H14O2.C5H8O2/c1-7(2)8(10)11-6-5-9(3)4;1-4-5-6-10-8(9)7(2)3;1-4(2)5(6)7-3/h1,5-6H2,2-4H3;2,4-6H2,1,3H3;1H2,2-3H3

InChI key

NEDGUIRITORSKL-UHFFFAOYSA-N

General description

Find answers to frequently asked questions on USP products in USP FAQs.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Amino methacrylate copolymer USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.


Storage Class

11 - Combustible Solids

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Skin Sens. 1 - STOT SE 3

target_organs

Respiratory system

flash_point_f

>482.0 °F

flash_point_c

> 250 °C



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library