Y0002169
Nomegestrol acetate impurity B
CRS, European Pharmacopoeia (EP) Reference Standard
Synonym(s):
Nomegestrol, 17-Hydroxy-6-methyl-19-norpregna-4,6-diene-3,20-dione
About This Item
Recommended Products
API family
megesterol
manufacturer/tradename
EDQM
storage temp.
2-8°C
SMILES string
O[C@]1([C@@]2([C@H]([C@H]3[C@@H]([C@H]4CCC(=O)C=C4C(=C3)C)CC2)CC1)C)C(=O)C
InChI
1S/C21H28O3/c1-12-10-18-16(15-5-4-14(23)11-17(12)15)6-8-20(3)19(18)7-9-21(20,24)13(2)22/h10-11,15-16,18-19,24H,4-9H2,1-3H3/t15-,16-,18-,19+,20+,21+/m1/s1
InChI key
KZUIYQJTUIACIG-YBZCJVABSA-N
General description
Application
Packaging
Other Notes
related product
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service