About This Item
Recommended Products
grade
primary reference standard
product line
phyproof® Reference Substance
Assay
≥75.0% (HPLC)
form
solid
manufacturer/tradename
PhytoLab
storage temp.
−20°C
SMILES string
O1COc2c1c(c3c(c2)[C@H]([C@@H]5[C@H](COC5=O)C3)c4cc(c(c(c4)OC)O)OC)O
InChI
1S/C21H20O8/c1-25-13-4-9(5-14(26-2)19(13)23)16-11-6-15-20(29-8-28-15)18(22)12(11)3-10-7-27-21(24)17(10)16/h4-6,10,16-17,22-23H,3,7-8H2,1-2H3/t10-,16+,17-/m0/s1
InChI key
JGGWNGRBXJWAOC-HKJPBSJPSA-N
Related Categories
General description
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 3
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service