Skip to Content

AR2016

Soy Bean Meal (N, S)

analytical standard

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.24
UNSPSC Code:
41116107
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

manufacturer/tradename

Alpha Resources

application(s)

food and beverages

format

matrix material

General description

Soy Bean Meal (N, S) is a reference standard produced in accordance to ISO Guide 34-2009. This reference has an indefinite known shelf life unopened. Once opened the shelf life is limited to two years. Keep sealed and store under normal dry laboratory conditions. This is a certified reference material (Working Standard) and is traceable to the above-mentioned reference standard(s).

Other Notes

Certified for the analytes listed below. See certificate for values and more details
Elemental Analysis: Nitrogen and Sulphur

Matrix Group: Nuts, Soy, Edible Oils and Fats


Storage Class

13 - Non Combustible Solids

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library