Skip to Content
Merck
All Photos(1)

Key Documents

80023458

TSKgel® Amide-80 (2 µm) HPLC Columns

L x I.D. 10 cm × 3.0 mm, carbamoyl phase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115709
L × I.D.:
10 cm × 3.0 mm
surface area:
450 m2/g
particle size:
2 μm
matrix active group:
carbamoyl phase
matrix:
fully porous particle

Product Name

TSKgel® Amide 80 HPLC Column, phase carbamoyl, L × I.D. 10 cm × 3.0 mm, 2 μm particle size

material

stainless steel

Agency

suitable for USP L68

product line

TSKgel®

manufacturer/tradename

Tosoh 23458

parameter

50 °C max. temp.
600 bar max. pressure

technique(s)

HPLC: suitable

L × I.D.

10 cm × 3.0 mm

surface area

450 m2/g

matrix

fully porous particle

matrix active group

carbamoyl phase

particle size

2 μm

operating pH range

2.0-7.5

separation technique

hydrophilic interaction (HILIC)

Looking for similar products? Visit Product Comparison Guide

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service