Skip to Content
Merck
All Photos(1)

Key Documents

5745-U

Supelco

ORBO 5030 (OVS) Amberlite XAD-7 Tube

pkg of 10 ea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
13111064
NACRES:
NB.24

material

RC,GFF,F,F (specially cleaned) separators

Quality Level

composition

Bed A, 250 mg
Bed B, 150 mg

packaging

pkg of 10 ea

O.D. × L

13-8 mm × 75 mm , OSHA Versatile Sampler, OVS

matrix

Amberlite XAD-7 (purified)

suitability

suitable for air monitoring
suitable for application (nonylphenol in the workplace)

application(s)

environmental
industrial hygiene

General description

ORBO 5030 (OVS) Amberlite XAD-7 Tube contains dual-layered beds of the same selective adsorbent separated by glass wool or foam, for gas and vapor sampling. This construction of the tube allows for any sample breakthrough to be captured in the smaller backup bed.

It is commonly used for acrylamide air sampling and analysis of its levels using HPLC-MSMS and for collecting propylene glycol and glycerol from air samples for subsequent analysis by GC-MS.

Application

ORBO 5030 (OVS) Amberlite XAD-7 tube has been used as an adsorbent for the collection and analysis of air sampling of trimethylamine (TMA) and dimethylamine (DMA) in confined atmospheres to capture the volatile amines.

Legal Information

Amberlite is a trademark of DuPont de Nemours, Inc.
ORBO is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Comparison of the effects of three types of heating tobacco system and conventional cigarettes on indoor air quality.
Yoshihiro E, et al.
SN Applied Sciences, 04, 1-19 (2022)
Optical fibre-based micro-analyser for indirect measurements of volatile amines levels in fish
Lurdes S BI, et al.
Food Chemistry, 123, 806-813 (2010)
Measurement of evaporated acrylamide during heat treatment of food and other biological materials.
Sune E, et al.
LWT--Food Science and Technology, 40, 706-712 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service