Skip to Content
Merck
All Photos(1)

Key Documents

42354

Supelco

Cereulide-13C6 (cyclo[-Val-1-13C-D-O-Leu-D-Ala-3-13C-O-Val-]3) solution

10 μg/g in acetonitrile, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

cyclo[-Val-1-13C-D-O-Leu-D-Ala-3-13C-O-Val-]3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C6C51H96N6O18
Molecular Weight:
1159.36
UNSPSC Code:
41116105
NACRES:
NA.21

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

concentration

10 μg/g in acetonitrile

format

single component solution

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma aldrich.com

The cyclic peptide Cereulide is a strong cytotoxin produced by some strains of Bacillus cereus and Bacillus megaterium. When ingested, it causes nausea and vomiting. It is both heat and acid stable and can be present mainly in stark-rich foods such as pasta or rice. This isotope-labelled certified reference material together with the native CRM solution 38708 is suitable to be used for ISO method 18465:2017 or similar LC-MS methods.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service