Documents
27375
Supelco
Crimp seals with PTFE/silicone septa
silver aluminum seal, seal diam. 20 mm, open center, white PTFE/silicone septum, pkg of 1000 ea
About This Item
UNSPSC Code:
41121821
material
silver aluminum seal
white PTFE/silicone septum
feature
open center
packaging
pkg of 1000 ea
liner thickness
5 mil
seal diam.
20 mm
compatibility
for use with 20 mm crimp top vial
Choose from one of the most recent versions:
Certificates of Analysis (COA)