ZRB1889
Anti-PTPN21 Antibody, clone 2K11 ZooMAb® Rabbit Monoclonal

recombinant, expressed in HEK 293 cells
Synonym(s):
Tyrosine-protein phosphatase non-receptor type 21;EC:3.1.3.48;Protein-tyrosine phosphatase D1;PTPD1
About This Item
Recommended Products
biological source
rabbit
Quality Level
recombinant
expressed in HEK 293 cells
conjugate
unconjugated
antibody form
purified antibody
antibody product type
primary antibodies
clone
2K11, recombinant monoclonal
description
2K11 Clone
product line
ZooMAb® learn more
form
lyophilized
mol wt
calculated mol wt 133.28 kDa
observed mol wt ~130 kDa
purified by
using Protein A
species reactivity
rat, mouse, human
packaging
antibody small pack of 25 μL
greener alternative product characteristics
Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.
enhanced validation
recombinant expression
Learn more about Antibody Enhanced Validation
sustainability
Greener Alternative Product
technique(s)
affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable
isotype
IgG
epitope sequence
C-terminal half
Protein ID accession no.
UniProt accession no.
greener alternative category
, Aligned
shipped in
ambient
storage temp.
2-8°C
target post-translational modification
unmodified
Gene Information
human ... PTPN21(11099)
General description
Specificity
Immunogen
Application
Evaluated by Western Blotting in ACHN cell lysate.
Western Blotting Analysis: A 1:1,000 dilution of this antibody detected PTPN21 in ACHN cell lysate.
Tested applications
Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected PTPN21 in A549, PC12, and Raw264.7 cell lysates.
Affinity Binding Assay: A representative lot of this antibody bound Ptpn21 peptide with a KD of 1.3 x 10-6 in an affinity binding assay.
Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected PTPN21 in human gallbladder tissue sections.
Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected PTPN21 in A549 cella.
Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Evaluated by Western Blotting in ACHN cell lysate.
Western Blotting Analysis: A 1:1,000 dilution of this antibody detected PTPN21 in ACHN cell lysate.
Target description
Physical form
Storage and Stability
Legal Information
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service