Key Documents
Z719471
Sorenson low binding aerosol barrier tips
MultiGuard, with graduation lines, maximum volume 200 μL
Synonym(s):
aerosol barrier tips, barrier filter pipette tip, barrier filter tips, barrier tip, filter pipette tip, filter tip, low bind tip, pipet tip
About This Item
UNSPSC Code:
41121601
NACRES:
NB.01
manufacturer/tradename:
Sorenson 17370T
maximum volume:
200 μL
sterility:
sterile
sterile
feature:
DNase free
RNase free
barrier
packaging:
pack of 96 ea (Racked)
material
colorless
natural tip (low binding)
polypropylene
sterility
sterile
sterile
feature
DNase free
RNase free
barrier
packaging
pack of 96 ea (Racked)
manufacturer/tradename
Sorenson 17370T
maximum volume
200 μL
binding type
low binding surface
General description
Tips feature a new surface technology designed for quantitative assays in which sample binding can significantly alter results or for critical small volume protocols that require a high degree of accuracy. Ideal for molecular biology applications, such as PCR. Inert hydrophobic barrier prevents pipettor contamination.
Features and Benefits
  • Suitable for DNA amplification and radioisotope handling
  • Over-pipetting does not contaminate valuable samples
  • Universal fit on research-grade pipettors
  • Filter devoid of leachable additives
  • Small micron pore sizes, <20 μ
  • Hydrophobic surface to lower surface tension
  • Minimal sample binding
  • Enhanced accuracy and precision
  • Additives-free such as silicone
  • Eliminates leaching and subsequent sample degradation
Legal Information
Sorenson is a trademark of Sorenson BioScience, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)