Z330868
Lenz® Squibb separating funnel
funnel capacity 100 mL, joint: ST/NS 19/26, glass key, graduated: no
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
material
borosilicate glass 3.3
glass key
Joint
joint: ST/NS19/26
feature
graduated: no
manufacturer/tradename
Bistabil®
Lenz 4.0100.37
funnel capacity
100 mL
plug bore
2.5 mm
Looking for similar products? Visit Product Comparison Guide
Legal Information
Bistabil is a registered trademark of Lenz GmbH
Lenz is a registered trademark of Lenz GmbH
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service