Skip to Content
Merck
All Photos(3)

Key Documents

WH0001774M2

Sigma-Aldrich

Monoclonal Anti-DNASE1L1 antibody produced in mouse

clone 4E8, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-DNAS1L1, Anti-DNL1L, Anti-XIB, Anti-deoxyribonuclease I-like 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4E8, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG1κ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DNASE1L1(1774)

General description

This gene encodes a member of the deoxyribonuclease family and the protein shows high sequence similarity to lysosomal DNase I. Alternate transcriptional splice variants, encoding the same protein, have been characterized. (provided by RefSeq)

Immunogen

DNASE1L1 (AAH01561, 18 a.a. ~ 302 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
AFRICAFNAQRLTLAKVAREQVMDTLVRILARCDIMVLQEVVDSSGSAIPLLLRELNRFDGSGPYSTLSSPQLGRSTYMETYVYFYRSHKTQVLSSYVYNDEDDVFAREPFVAQFSLPSNVLPSLVLVPLHTTPKAVEKELNALYDVFLEVSQHWQSKDVILLGDFNADCASLTKKRLDKLELRTEPGFHWVIADGEDTTVRASTHCTYDRVVLHGERCRSLLHTAAAFDFPTSFQLTEEEALNISDHYPVEVELKLSQAHSVQPLSLTVLLLLSLLSPQLCPAA

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tingting Wang et al.
Frontiers in immunology, 10, 1235-1235 (2019-06-20)
Immune checkpoint blockade (ICB) immunotherapy increases antitumor immunity by blocking cytotoxic-T-lymphocyte-associated protein 4 (CTLA-4) or programmed cell death protein 1 (PD-1)/programmed death-ligand 1 (PD-L1) and displays robust clinical responses in various cancers. However, ICB immunotherapy also triggers severe inflammatory side
Katarzyna Biadasiewicz et al.
Endocrinology, 152(4), 1458-1469 (2011-02-10)
The basic helix-span-helix transcription factor activating protein (AP)-2α is critically involved in cell-specific hormone expression of syncytializing human trophoblasts. Its role in invasive trophoblast differentiation, however, remains largely elusive. Using RT-PCR, Western blotting, and immunofluorescence of first-trimester placentae, we here

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service