Merck
T3418
Nalgene® centrifuge tubes, Oak Ridge Style 3138
nominal capacity 50 mL
Synonym(s):
High-Speed ​​Centrifuge Tubes, Nalgene 3138-0050, sample collection tube, sample receptacle, sample tube
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
41121703
NACRES:
NB.11
eCl@ss:
32119210
Material:
cap (PP sealing cap), round bottom tube, transparent polycarbonate
Sterility:
non-sterile
Packaging:
pack of 10 ea
Feature:
RCF 50,000 × g
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
material
cap (PP sealing cap), round bottom tube, transparent polycarbonate
sterility
non-sterile
feature
RCF 50,000 × g
packaging
pack of 10 ea
manufacturer/tradename
Nalgene® 3138-0050
nominal capacity
50 mL
General description
Easily view contents using strong, transparent Thermo Scientific™ Nalgene™ Oak Ridge High-Speed Polycarbonate Centrifuge Tubes, equipped with sealing caps for leakproof† performance up to 50,000 x g.
Packaging
With sealing cap
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Still not finding the right product?
Choose from one of the most recent versions:
Certificates of Analysis (COA)