Skip to Content
Merck

SML3586

MU1700 dihydrochloride

≥98% (HPLC)

Synonym(s):

6-(4-(Piperazin-1-yl)phenyl)-3-(quinolin-4-yl)furo[3,2-b]pyridine dihydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C26H22N4O ·2HCl
Molecular Weight:
479.40
NACRES:
NA.21
UNSPSC Code:
12352200
Assay:
≥98% (HPLC)
Form:
powder
Storage condition:
desiccated
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear (Warmed)

storage temp.

2-8°C

Biochem/physiol Actions

MU1700 is a potent and selective TGF-β type I receptors ALK1/ACVRL1 & ALK2/ACVR1 inhibitor (IC50 = 13/6 nM, [ATP] = 10 mM) that inhibits ALK6/BMPR1B only at higher concentrations (IC50 = 41 nM) with good kinome selectivity over >360 other kinases, including ALK3/BMPR1A (IC50 = 425 nM), ALK4/ACVR1B & ALK5/TGFBR1 (inactive). MU1700 inhibits cellular phospho-SMAD1/5/8 induction by 10 ng/mL BMP-7 and phospho-SMAD2 induction by either 10 ng/mL activin A or 100 ng/mL TGF-β1 with good target selectivity by cellular NanoBRET assays (ALK1/2 IC50 = 225/27 nM).
Potent and selective TGF-β type I receptors ALK1/ACVRL1 & ALK2/ACVR1 inhibitor.

Disclaimer

Hygroscopic


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library