Skip to Content
Merck
All Photos(1)

Key Documents

SML3389

Sigma-Aldrich

Vilanterol trifenatate

≥95% (HPLC)

Synonym(s):

4-[(1R)-2-({6-[(2-{[(2,6-Dichlorophenyl)methyl]oxy}ethyl)oxy]-hexyl}-amino)-1-hydroxyethyl]-2-(hydroxymethyl)phenol, triphenylacetate salt, GW 642444 trifenatate, GW 642444M, GW-642444 trifenatate, GW-642444M, GW642444 trifenatate, GW642444M, Vilanterol triphenylacetate sal

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H33Cl2NO5·C20H16O2
CAS Number:
Molecular Weight:
774.77
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

OC(C(C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3)=O.ClC(C=CC=C4Cl)=C4COCCOCCCCCCNC[C@@H](C5=CC(CO)=C(C=C5)O)O

InChI

1S/C24H33Cl2NO5.C20H16O2/c25-21-6-5-7-22(26)20(21)17-32-13-12-31-11-4-2-1-3-10-27-15-24(30)18-8-9-23(29)19(14-18)16-28;21-19(22)20(16-10-4-1-5-11-16,17-12-6-2-7-13-17)18-14-8-3-9-15-18/h5-9,14,24,27-30H,1-4,10-13,15-17H2;1-15H,(H,21,22)/t24-;/m0./s1

InChI key

KLOLZALDXGTNQE-JIDHJSLPSA-N

General description

Vilanterol trifenatate is a potent and selective beta2 (β2)-adrenoceptor agonist. This bioactive small molecule has been shown to stimulate cellular cAMP accumulation in CHO cells that express human beta2 adrenoceptors, while not having the same effect on beta1 or beta3 receptors (pEC50 = 9.4/β2, 6.4/β1, 6.1/β3). Vilanterol can reduce the contraction of electrically stimulated guinea pig trachea strips and exhibits in vivo therapeutic efficacy in an asthma model in guinea pigs with histamine-induced bronchospasm (EC90 = nebulizer concentration 30 μM). Additionally, Vilanterol trifenatate acts as a substrate for P-glycoprotein (PgP).

Biochem/physiol Actions

Potent and selective, beta2 (β2)-adrenoceptor agonist in vitro and in vivo.
Vilanterol is a potent and selective, beta2 (β2)-adrenoceptor agonist that stimulates cellular cAMP accumulation in CHO cells expressing human beta2, but not beta1 or beta3, adrenoceptor (pEC50 = 9.4/β2, 6.4/β1, 6.1/β3). Vilanterol inhibits the contraction of electrically stimulated guinea pig trachea strips and displays in vivo therapeutic efficacy in a guinea pig asthma model of histamine-induced bronchospasm (EC90 = nebulizer concentration 30 μM).

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Skin Irrit. 2 - STOT SE 2

Target Organs

Cardiovascular

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mario Cazzola et al.
British journal of pharmacology, 163(1), 4-17 (2011-01-15)
Despite the passionate debate over the use of β(2) -adrenoceptor agonists in the treatment of airway disorders, these agents are still central in the symptomatic management of asthma and COPD. A variety of β(2) -adrenoceptor agonists with long half-lives, also
The effect of ketoconazole on the pharmacokinetics and pharmacodynamics of inhaled fluticasone furoate and vilanterol trifenatate in healthy subjects
Kempsford R, et al.
British Journal of Clinical Pharmacology (2012)
Panayiotis A Procopiou et al.
Journal of medicinal chemistry, 53(11), 4522-4530 (2010-05-14)
A series of saligenin beta(2) adrenoceptor agonist antedrugs having high clearance were prepared by reacting a protected saligenin oxazolidinone with protected hydroxyethoxyalkoxyalkyl bromides, followed by removal of the hydroxy-protecting group, alkylation, and final deprotection. The compounds were screened for beta(2)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service