Skip to Content
Merck
All Photos(1)

Key Documents

SML3268

Sigma-Aldrich

Vaccarin

≥98% (HPLC)

Synonym(s):

6-(2-O-α-L-Arabinopyranosyl-β-D-glucopyranosyl)-2-[4-(β-D-glucopyranosyloxy)phenyl]-5,7-dihydroxy-4H-1-benzopyran-4-one, CID 44257777

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H38O19
CAS Number:
Molecular Weight:
726.63
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (Warmed)

storage temp.

-10 to -25°C

Biochem/physiol Actions

Active ingredient of Semen Vaccaria; antidiabetic, endothelial cell injury prevention, angiogenesis promotion, wound healing, and liver protection
Vaccarin is a main active ingredient of medical plant Semen Vaccaria that exhibits varies biological activities including antidiabetic, endothelial cell injury prevention, angiogenesis promotion, wound healing, and liver protection. Vvaccarin protects the intestinal barrier by inhibiting the ERK/MLCK signaling pathway in type 2 diabetic mice.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bao Hou et al.
Acta cirurgica brasileira, 34(12), e201901202-e201901202 (2020-02-13)
To explore the potential role and unclear molecular mechanisms of vaccarin in wound healing. Rats' skin excision model to study the effects of vaccarin on wound healing in vivo . Hematoxylin and eosin staining was performed to evaluate Histopathologic characteristics.
Jiang-Nan Sun et al.
European journal of pharmacology, 908, 174375-174375 (2021-07-26)
Hyperglycemia and insulin resistance drive intestinal barrier dysfunction in type 2 diabetes (T2DM). Vaccarin, the main active component in the semen of traditional Chinese medicine Vaccaria has a definite effect on T2DM mice. The purpose of this study was to
Yixiao Liu et al.
International journal of molecular sciences, 21(6) (2020-03-19)
Diabetes mellitus is a growing global health issue nearly across the world. Diabetic patients who are prone to develop diabetes-related complications often exhibit progressive neuropathy (painless and sensory loss). It is usual for small wounds to progress to ulceration, which

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service