Skip to Content
Merck
All Photos(1)

Documents

SML2852

Sigma-Aldrich

Capadenoson

≥98% (HPLC)

Synonym(s):

2-Amino-6-[[[2-(4-chlorophenyl)-1,3-thiazol-4-yl]methyl]sulfanyl]-4-[4-(2-hydroxyethoxy)phenyl]pyridine-3,5-dicarbonitrile, 2-Amino-6-[[[2-(4-chlorophenyl)-4-thiazolyl]methyl]thio]-4-[4-(2-hydroxyethoxy)phenyl]-3,5-pyridinedicarbonitrile, BAY 68-4986, BAY-68-4986, BAY68-4986

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H18ClN5O2S2
CAS Number:
Molecular Weight:
520.03
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

N#CC1=C(C2=CC=C(OCCO)C=C2)C(C#N)=C(N)N=C1SCC3=CSC(C4=CC=C(Cl)C=C4)=N3

InChI

1S/C25H18ClN5O2S2/c26-17-5-1-16(2-6-17)24-30-18(13-34-24)14-35-25-21(12-28)22(20(11-27)23(29)31-25)15-3-7-19(8-4-15)33-10-9-32/h1-8,13,32H,9-10,14H2,(H2,29,31)

InChI key

CITWCLNVRIKQAF-UHFFFAOYSA-N

Biochem/physiol Actions

Capadenoson (BAY 68-4986) is an adenosine receptor A1 (A1AR) partial agonist (GTPγS binding EC50/Emax = 0.1 nM/74% vs. 0.3 nM/100% with CCPA; human cortex membranes) that exerts additional biased A2BAR agonism toward cAMP signal transduction (pEC50 = 8.94/cAMP, 6.20/Ca+2, 6.12/pERK, 5.03/IP1), while exhibiting weak A2A & A3 potency. Capadenoson offers cardioprotection efficacy in an ischemia-reperfusion injury rat model in vivo (25/28% infarct size reduction with 0.1/0.3 mg/kg iv.) without the risk of a full atrioventricular (AV) block (isolated perfused rat heart rate = 100% up to 10 nM, 90% at ≥10 μM) seen with the full A1 agonist CCPA.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniel Meibom et al.
ChemMedChem, 12(10), 728-737 (2017-05-11)
Adenosine is known to be released under a variety of physiological and pathophysiological conditions to facilitate the protection and regeneration of injured ischemic tissues. The activation of myocardial adenosine A1 receptors (A1 Rs) has been shown to inhibit myocardial pathologies associated
Barbara E Albrecht-Küpper et al.
Purinergic signalling, 8(Suppl 1), 91-99 (2011-11-15)
Adenosine, a purine nucleoside, is present in all cells in tightly regulated concentrations. It has many different physiological effects in the whole body and in the heart. Adenosine activates four G protein-coupled receptors A1, A2a, A2b, and A3. Activation of
Isaac R Bailey et al.
The Journal of pharmacology and experimental therapeutics, 362(3), 424-430 (2017-06-28)
Cardiac arrest is a leading cause of death in the United States, and, currently, therapeutic hypothermia, now called targeted temperature management (TTM), is the only recent treatment modality proven to increase survival rates and reduce morbidity for this condition. Shivering
Jo-Anne Baltos et al.
Biochemical pharmacology, 135, 79-89 (2017-03-28)
The adenosine A2B receptor (A2BAR) has been identified as an important therapeutic target in cardiovascular disease, however in vitro and in vivo targeting has been limited by the paucity of pharmacological tools, particularly potent agonists. Interestingly, 2-((6-amino-3,5-dicyano-4-(4-(cyclopropylmethoxy)phenyl)-2-pyridinyl)thio)acetamide (BAY60-6583), a potent
Samantha L Cooper et al.
British journal of pharmacology, 177(2), 346-359 (2019-10-10)
Adenosine is a local mediator that regulates physiological and pathological processes via activation of four GPCRs (A1 , A2A , A2B , and A3 ). We have investigated the effect of two A1 -receptor-selective agonists and the novel A1 -receptor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service