Skip to Content
Merck
All Photos(1)

Key Documents

S1509

Supelco

Sigma TLC spray box

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115711
NACRES:
NC.07

technique(s)

thin layer chromatography (TLC): suitable

size

~14 in. × 14 in. × 14 in.

General description

Completely disposable.
Contains knock out vents for better ventilation. Easy to assemble.
The reagent/chemical spraying on the TLC plates are generally carried out inside a fume hood or commercially available TLC spray cabinet with blower and exhaust hose. The TLC plate can be placed supported upright or on a paper inside the spray box. Spraying the box ensures complete removal of reagent mist.

Application

  • Sigma TLC spray box may be used to spray reagent for TLC plates.
  • Sigma TLC spray box may be used in thin layer chromatography and HPTLC.

Compatibility

Compatible with most Sigma sprays.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Detection (visualization) of TLC zones.
Sherma, Joseph
Encyclopedia of Chromatography, 1, 449-455 (2005)
Yu-Jiao Cai et al.
PloS one, 8(3), e58647-e58647 (2013-04-05)
Epithelial cells(EC)-derived interleukin-7 (IL-7) plays a crucial role in control of development and homeostasis of neighboring intraepithelial lymphocytes (IEL), and keratinocyte growth factor (KGF) exerts protective effects on intestinal epithelial cells and up-regulates EC-derived IL-7 expression through KGFR pathway. This

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service