Skip to Content
Merck

MTCT7950

MTC Bio Sample Transport Tubes

capacity 14 mL, self-standing

Synonym(s):

reagent dish, sample storage tube, self standing tube, MTC Bio T9005

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NE.25
UNSPSC Code:
41121706
Capacity:
14 mL
Material:
clear , polypropylene , screw cap
Sterility:
sterile
Packaging:
pack of 1000 ea (10 x bags 100 ea)
Feature:
graduation
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

clear , polypropylene , screw cap

sterility

sterile

feature

graduation

packaging

pack of 1000 ea (10 x bags 100 ea)

manufacturer/tradename

MTC Bio T7950

capacity

14 mL

diam. × L

16 mm × 100 mm

suitability

suitable for (cell culture)

General description

16 x 100 mm screw cap test tubes are available in transparent polypropylene with a “skirted” self-standing bottom or in crystal clear polystyrene with a traditional round bottom. Each tube has easy to read graduations and a “frosted” white marking area (12x50mm). Packed 100 tubes (with attached caps) per sterile bag, 10 bags per unit.

Important: Caps may be supplied loosely attached to allow effective sterilization. Product sterility is maintained by the sealed packaging. Once the sterile bag is opened, sterility of any unused tubes is not guaranteed.
Transport/ Culture Tube, 16 x 100mm, PP, self-standing, with screw-cap, white printed graduations, 14mL, sterile, 10 bags of 100 tubes, 1000/cs

Important: Caps may be supplied loosely attached to allow effective sterilization. Product sterility is maintained by the sealed packaging. Once the sterile bag is opened, sterility of any unused tubes is not guaranteed.

Features and Benefits

  • Leak-proof screw caps
  • White printed graduations
  • Sterile bags of 100 tubes
  • White marking spot
  • Self-standing / round bottom

Legal Information

MTC is a trademark of MTC Bio


Still not finding the right product?

Explore all of our products under MTC Bio Sample Transport Tubes




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library