MSST0035
SILu™Prot CCL2 C-C motif chemokine 2 human
SIL MS Protein Standard, 13C and 15N-labeled
Synonym(s):
HC11, Monocyte chemoattractant protein 1, Monocyte chemotactic protein 1(MCP-1), Monocyte secretory protein JE, Monocytechemoattractantprotein1, Monocytechemotacticandactivatingfactor(MCAF), Small-inducible cytokine A2
About This Item
Recommended Products
biological source
human
Quality Level
Assay
≥98% (SDS-PAGE)
form
lyophilized powder
potency
≥98% Heavy amino acids incorporation efficiency by MS
technique(s)
mass spectrometry (MS): suitable
suitability
suitable for mass spectrometry (standard)
UniProt accession no.
storage temp.
−20°C
Gene Information
human ... CCL2(6347)
General description
Immunogen
Biochem/physiol Actions
Physical form
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service