M3311
Greiner 384 well plates, polystyrene
working volume 110 μL, flat bottom clear polystyrene wells
Synonym(s):
384 multiwell plates, 384 well microplates, 384 well microtiter plates, 384 well plates
About This Item
Recommended Products
material
clear polystyrene plate
flat bottom clear polystyrene wells
polystyrene
sterility
non-sterile
feature
lid: no
packaging
case of 100
manufacturer/tradename
Greiner 781101
maximum volume
130 μL
size
384 wells
working volume
110 μL
color
clear
binding type
non-treated surface
Looking for similar products? Visit Product Comparison Guide
General description
Other Notes
Suitability
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service