Skip to Content
Merck

L7784

Lewis-Y tetrasaccharide

Synonym(s):

α-Fuc-(1→2)-β-Gal-(1→4)-(α-Fuc-[1→3])-GlcNAc

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C26H45NO19
Molecular Weight:
675.63
NACRES:
NA.32
PubChem Substance ID:
UNSPSC Code:
12352202
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


storage temp.

2-8°C

Quality Segment

SMILES string

C[C@H]1O[C@H](O[C@@H]2[C@@H](O)[C@@H](O)[C@@H](CO)O[C@H]2O[C@@H]3[C@@H](CO)OC(O)[C@H](NC(C)=O)[C@H]3O[C@H]4O[C@H](C)[C@H](O)[C@H](O)[C@H]4O)[C@H](O)[C@@H](O)[C@H]1O

InChI

1S/C26H45NO19/c1-6-12(31)15(34)18(37)24(40-6)45-21-11(27-8(3)30)23(39)42-10(5-29)20(21)44-26-22(17(36)14(33)9(4-28)43-26)46-25-19(38)16(35)13(32)7(2)41-25/h6-7,9-26,28-29,31-39H,4-5H2,1-3H3,(H,27,30)/t6-,7-,9-,10-,11-,12+,13+,14+,15+,16+,17+,18-,19-,20-,21-,22-,23?,24-,25-,26+/m1/s1

InChI key

SRHNADOZAAWYLV-MVSAILQPSA-N

Application

Lewis-Y blood group antigenic determinant.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library