L7771
L-(+)-Lactic acid silver salt
~90%
Synonym(s):
2-Hydroxypropionic acid silver salt, Sarcolactic acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C3H5AgO3
CAS Number:
Molecular Weight:
196.94
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Recommended Products
Assay
~90%
SMILES string
CC1O[Ag]OC1=O
InChI
1S/C3H5O3.Ag/c1-2(4)3(5)6;/h2H,1H3,(H,5,6);/q-1;+2/p-1
InChI key
HCSWQVIOAPZWTM-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
Application
Used for light microscopic visualization of colloidal gold in tissue.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Ayten Azizova Gurbanova et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 28(31), 7828-7836 (2008-08-01)
We showed previously that genetic absence epilepsy rats from Strasbourg (GAERS) resist secondary generalization of focal limbic seizures after electrical kindling. We now investigate the effect of intra-amygdaloid injection of kainic acid, as another model of temporal lobe epilepsy, focusing
G Danscher et al.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society, 31(12), 1394-1398 (1983-12-01)
RNase labeled with colloidal gold was used as a model for the present technique evolved for the light microscopic localization of gold-labeled substances in semithin resin-embedded sections. Tissue sections placed on glass slides were treated with the gold-enzyme complex and
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service