Skip to Content
Merck
All Photos(1)

Key Documents

JN0003

Sigma-Aldrich

Ridogrel

≥98% (HPLC)

Synonym(s):

5-[(E)-[3-pyridyl-[3-(trifluoromethyl)phenyl]methylene]amino]oxypentanoic acid, R 68070, R068070

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H17F3N2O3
CAS Number:
Molecular Weight:
366.33
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

room temp

SMILES string

O=C(O)CCCCO/N=C(C1=CC=CN=C1)\C2=CC=CC(C(F)(F)F)=C2

InChI

1S/C18H17F3N2O3/c19-18(20,21)15-7-3-5-13(11-15)17(14-6-4-9-22-12-14)23-26-10-2-1-8-16(24)25/h3-7,9,11-12H,1-2,8,10H2,(H,24,25)/b23-17-

InChI key

GLLPUTYLZIKEGF-QJOMJCCJSA-N

Biochem/physiol Actions

A2 receptor antagonist
Ridogrel is an orally active, potent and specific combined thromboxane synthase inhibitor and thromboxane A2 receptor ( thromboxane/prostaglandin endoperoxide receptor) antagonist. Ridogrel is a potent antiplatelet agent.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E Carty et al.
Alimentary pharmacology & therapeutics, 14(6), 807-817 (2000-06-10)
Thromboxanes, prostaglandins, reactive oxygen metabolites and pro-inflammatory cytokines are produced in excess in inflammatory bowel disease. Preliminary reports suggest that ridogrel, a thromboxane synthesis inhibitor and receptor blocker, may have therapeutic benefits in ulcerative colitis. To investigate the anti-inflammatory profile
J J Auwerda et al.
European journal of gastroenterology & hepatology, 13(4), 397-400 (2001-05-08)
To evaluate the effect of Ridogrel enemas (Janssen Research Foundation, Beerse, Belgium) on disease activity and mucosal inflammatory mediators in patients with active left-sided ulcerative colitis. Eleven patients with active left-sided ulcerative colitis were evaluated in an open non-placebo-controlled pilot

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service