Skip to Content
Merck
All Photos(4)

Key Documents

HPA024745

Sigma-Aldrich

Anti-VPS28 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2

Synonym(s):

Anti-ESCRT-I complex subunit VPS28, Anti-H-Vps28, Anti-Vacuolar protein sorting-associated protein 28 homolog

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

independent
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

VQYKAAFRQVQGSEISSIDEFCRKFRLDCPLAMERIKEDRPITIKDDKGNLNRCIADVVSLFITVMDKLRLEIR

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... VPS28(51160)

General description

The gene VPS28 (vacuolar protein sorting-associated protein 28 homolog) is mapped to human chromosome 8q24.3. The encoded protein is part of the ESCRT-1 (endosomal complexes required for transport) complex. The complex participates in membrane remodelling, including multivesicular body biogenesis and cytokinesis.

Immunogen

Vacuolar protein sorting-associated protein 28 homolog recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

VPS28 (vacuolar protein sorting-associated protein 28 homolog) participates in various cellular events, including OGD/R (oxygen and glucose deprivation followed by reoxygenation)-mediated neuronal cell death, peptide growth factors-initiated SOS1 (son of sevenless 1)-Ras signaling, epithelial-mesenchymal transition and cell invasion.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST76323

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Hawthorn et al.
Genomics, 103(2-3), 211-221 (2013-12-10)
We compared transcript expression, and chromosomal changes on a series of tumors and surrounding tissues to determine if there is evidence of field cancerization in colorectal cancer. Epithelial cells were isolated from tumors and areas adjacent to the tumors ranging
Sun-Young Han et al.
Biochemical and biophysical research communications, 387(3), 548-552 (2009-07-21)
Epithelial-mesenchymal transition (EMT) and the acquisition of invasive potential are key events in tumor progression. We now show that CIIA, originally identified as an anti-apoptotic protein, induces the EMT and promotes cell migration and invasion. Ectopic expression of CIIA induced
Hyun Sub Hwang et al.
Journal of cell science, 127(Pt 8), 1640-1646 (2014-02-14)
Son of sevenless 1 (SOS1) is a Ras-specific guanine-nucleotide-exchange factor (GEF) that mediates intracellular signaling processes induced by receptor tyrosine kinases. In this study, we show that CIIA (also known as VPS28) physically associates with SOS1 and thereby inhibits the
Sang Gil Hwang et al.
Molecular and cellular biochemistry, 397(1-2), 139-146 (2014-08-08)
Brain ischemia causes neuronal injury leading to stroke and other related brain diseases. However, the precise mechanism of the ischemia-induced neuronal death remains unclear yet. In this study, we showed that CIIA suppressed neuronal cell death induced by oxygen and
Giancarlo O Tanzi et al.
Journal of virology, 77(15), 8440-8447 (2003-07-15)
A final step in retrovirus assembly, particle release from the cell, is modulated by a small motif in the Gag protein known as a late domain. Recently, human immunodeficiency virus type 1 (HIV-1) and Moloney murine leukemia virus (M-MuLV) were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service