Skip to Content
Merck
All Photos(1)

Key Documents

GERPN5785

Vistra ECF Substrate

Cytiva RPN5785

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105332
NACRES:
NA.32

packaging

pkg of 1 kit

manufacturer/tradename

Cytiva RPN5785

color

ECF substrate: Off-white/white to yellowish ECF substrate dilution buffer: Colorless

shipped in

wet ice

storage temp.

2-8°C

General description

Alkaline phosphatase dephosphorylates ECF Substrate to produce a fluorescent product, which localizes at the site of catalysis. Product fluoresces at 540 to 560 nm when excited at the appropriate wavelength.

The following reagents are provided, sufficient for detecting 2500 cm2 membrane: ECF Substrate, 60 mL, and dilution buffer.

Application

Vistra ECF substrate has been used in Western blotting.

Features and Benefits

  • A fluorescent substrate for alkaline phosphatase-based detection of protein blots, for use with fluorescence scanning equipment such as Storm instruments (see Storm)
  • For use in western blotting, dot, and slot blotting applications
  • Sensitivity comparable to chemiluminescence
  • The wider dynamic range of fluorescence scanning systems compared with film allows visualization and quantitation of weak and dark bands in the same scan
  • Eliminates need for darkroom and film

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

ECF is a trademark of Cytiva

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Storage Class Code

12 - Non Combustible Liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anti-atherosclerotic effects of small molecular weight compounds enhancing eNOS expression and preventing eNOS uncoupling.
Wohlfart P, et al.
Journal of Pharmacology and Experimental Therapeutics (2007)
Cindy Gueguen et al.
PloS one, 9(2), e87367-e87367 (2014-03-04)
Poly(ADP-ribose)polymerase and sirtuin 1 are both NAD(+)-dependent enzymes. In vitro oxidative stress activates poly(ADP-ribose)polymerase, decreases NAD(+) level, sirtuin 1 activity and finally leads to cell death. Poly(ADP-ribose)polymerase hyperactivation contributes to cell death. In addition, poly(ADP-ribose)polymerase inhibition restores NAD(+) level and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service