Skip to Content
Merck
All Photos(1)

Key Documents

GE17-1087-01

SP Sepharose High Performance

Cytiva 17-1087-01, pack of 75 mL

Synonym(s):

SP Sepharose

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106500
NACRES:
NA.56

ligand

sulphopropyl

description

Ion Exchanger Type (value)

packaging

pack of 75 mL

manufacturer/tradename

Cytiva 17-1087-01

matrix

6% cross-linked agarose

particle size

24-44 μm

avg. part. size

34 μm

cleaning in place

3-14

working range

4-13

suitability

suitable for bioprocess medium

General description

SP Sepharose High Performance is a strong cation exchange BioProcess resin with high resolution designed for intermediate and polishing steps in downstream processing. SP Sepharose High Performance is composed of crosslinked agarose beads with a mean diameter of 34 µm, modified with sulphopropyl (SP) strong cation exchange groups. It is designed for intermediate purification and polishing applications and to be used when resolution and capacity have priority. SP Sepharose High Performance has high chemical stability and withstands effective CIP/sanitization protocols using sodium hydroxide.

Features and Benefits

  • Strong cation exchanger for intermediate and final polishing steps
  • High resolution, high capacity separation with high recovery
  • High chemical stability for effective CIP/sanitization
  • BioProcess resin supported for industrial applications and well-established in approved processes

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Store at 4 to 30 °C (20% Ethanol + 0.2 M Sodium Acetate)

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

Sepharose is a trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

This page describes principles and standard conditions for different purification techniques of histidine-tagged proteins using Cytiva products.

Protocols

This page covers detailed information on cleaning procedures and recommended flow for column cleaning.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service