Key Documents
EP0030131525
Eppendorf® plate lid
sterile, pkg of 80 ea
About This Item
UNSPSC Code:
41121800
NACRES:
NB.22
sterility
sterile
packaging
pkg of 80 ea
manufacturer/tradename
Eppendorf® 0030131525
Features and Benefits
  • Sealing Mats are reusables and can be autoclave. Sealing mats are suitable for short-term storage
    • Heat Sealing Film and Foil for continuous locking give the best evaporation protection
    • Foil products are made of aluminum, can be pierced, and protect light-sensitive samples
    • Film products are transparent and provide protection against unwanted punctures
  • Eppendorf Plate Lid: stable, flexible protection of samples during short-term storage
    • Storage Film and Foil glued seals for simple and dependable sealing during sample storage
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)