Skip to Content
Merck
All Photos(1)

Key Documents

EP0030122356

Eppendorf® Protein LoBind Tubes

capacity 5 mL, screw cap, PCR clean, pack of 200 ea (2 bags of 100 Tubes each)

Synonym(s):

Eppendorf LoBind, LoBind tubes

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121703
NACRES:
NB.11

material

conical bottom
polypropylene tube
white polypropylene cap

sterility

non-sterile

feature

PCR clean
Eppendorf Tube®
screw cap

packaging

pack of 200 ea (2 bags of 100 Tubes each)

capacity

5 mL

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

Application

  • Preparation or storage of protein, peptide or antibody samples.
  • Enzymatic assays – the hydrophilic surface reduces denaturation when it comes into contact with the tube inner wall.
  • Prevention of sample loss during storage of virus stock solutions.
  • Storage of cell suspensions.

Legal Information

Eppendorf is a registered trademark of Eppendorf AG
Eppendorf Tube is a registered trademark of Eppendorf AG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service