EP0030113560
Eppendorf twin.tec® PCR Plates 96
skirt, PCR clean, working volume 150 μL, colorless (with 2D SafeCode), pkg of 25 ea
Synonym(s):
Plates
About This Item
polypropylene
lid: no
skirt
Recommended Products
material
colorless (with 2D SafeCode)
polypropylene
sterility
non-sterile
feature
PCR clean
lid: no
skirt
packaging
pkg of 25 ea
manufacturer/tradename
Eppendorf® 0030113560
size
96 well
well size
150 μL , per well
working volume
150 μL
suitability
suitable for PCR
Looking for similar products? Visit Product Comparison Guide
General description
Application
Features and Benefits
- 3-level coding for safe sample and vessel identification
- Codes on three sides of plates for flexible reading
- ECC 200 error correction function enables safe code reading with up to 30 % damaged codes
- Additional, ID-specific information on plates available via Eppendorf DataPort for documentation purposes
- Direct import into eLabInventory and eLabJournal software for convenient handling
- Import to other database systems possible
- Manufactured from high-quality polypropylene for highest sample purity
- Maximum recovery rates for improved assay results
- Available with barcode (upon request)
- Ideal for quantitative real-time PCR as well
- Exceptionally solid and torque-resistant polycarbonate frame
- OptiTrack® matrix for faster sample identification and fewer pipetting errors
- Raised well rims for effective sealing, also reduces risk of cross-contamination
- Batch-tested and certified free of DNA, DNAase, RNase and PCR inhibitors (PCR clean)
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service