CLS8509
Corning® 384 2D Barcoded Storage Tubes
black polypropylene, round bottom, with generic barcode, non-sterile, lid: no, pack of 384
Synonym(s):
sample tube
About This Item
Recommended Products
material
black polypropylene
black wells
round bottom
sterility
non-sterile
feature
lid: no
packaging
pack of 384
case of 7680
manufacturer/tradename
Corning 8509
parameter
-80-110 °C temp. range
L
14 mm
capacity
120 μL
size
384 wells
tube H
16.9 mm , with cap
working volume
100 μL
color
black
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Features include:
- IDENTIFICATION - Synchronized 2D and linear bar code - unlike any other in the market
- INFORMATION - 14 x 14 dot 2D bar code
- BAR CODE STABILITY - Laser-etched, not an attached label
- COMPOUND COMPATIBILITY - Polypropylene construction throughout
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service